Antibody column package, Modern Protein A (1-step purification protocol)
Proveedor: Cytiva
29-4976-28
29-4976-28EA 267 EUR
29-4976-28
Antibody column package, Modern Protein A (1-step purification protocol)
Columnas de cromatografía
The Antibody column package includes modern Protein A affinity resin and desalting columns for purification of human antibodies.
  • Contains columns for a 1-step protocol using affinity chromatography, HiTrap MabSelect PrismA 1×1 ml and HiTrap Desalting 1×5 ml
  • HiTrap MabSelect PrismA column is prepacked with resin for preparative purification of monoclonal and polyclonal antibodies from serum and cell culture supernatant
  • HiTrap Desalting column is packed with Sephadex G-25 to give you quick pH neutralisation of the eluted antibodies
  • Includes instruction with