VWR®, Test Sieves, 300×60 mm, with Square Holes
Proveedor: VWR Collection
510-2789EA 281 EUR
510-2789 510-2790 510-2791 510-2792 510-2793 510-2794 510-2795 510-2796 510-2797 510-2798 510-2799 510-2801 510-2802 510-2803 510-2800 510-2804 510-2805 510-2806 510-2807 510-2808 510-2809 510-2810 510-2811 510-2812 510-2813 510-2814 510-2815 510-2816 510-2817 510-2818 510-2819 510-2820 510-2821 510-2822 510-2823 510-2824 510-2825 510-2826 510-2827 510-2828 510-2829
VWR®, Test Sieves, 300×60 mm, with Square Holes
Tamices Tamices de ensayo
Frame: Stainless steel; perforated plates: Galvanised steel, ISO 3310-2, ASTM E323, BS 410, AFNOR.
Optional: Inspection certificate of calibration 3.1 according to DIN EN 10204 with measuring results of a specific test.
Sieves and accessories of other dimensions are available on request.
Please go to website for a full overview.
All sieves are supplied with serial number and a certificate of compliance with the order 2.1 according to DIN EN 10204. Further certificates and calibration services are available. Contact your local Avantor sales office for further details.
InformaciónMore Product Information