Bromobimane (mBBr) ≥95%

Proveedor: AAT BIOQUEST
Atención

Sinónimos: Monobromobimane, 3-(Bromomethyl)-2,5,6-trimethylpyrazolo[1,2-a]pyrazole-1,7-dione

633
AATB633EA 120 EUR
AATB633
Bromobimane (mBBr) ≥95%
Bromobimane (mBBr)

mBBr is one of the most popular thiol-reactive fluorescent tags, and is widely used to detect various thiol-containing biomolecules such as glutathione in cells.

Formula: C₁₀H₁₁BrN₂O₂
Número MDL: MFCD00036951
Núm. CAS: 71418-44-5
ADR: 5,II

Order Now


Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Más About VWR