Magicard™ Colour + Black Film for Magicard™ 600, 250 Shot
Proveedor: Brady
321968
BRDY321968EA 104 EUR
BRDY321968
Magicard™ Colour + Black Film for Magicard™ 600, 250 Shot
Cintas de impresora
The Magicard™ combined colour/black ribbon is perfect for full colour prints on one side and black prints on the other side, using a Magicard™ 600 printer.
  • High-quality, full colour ribbon with overcoat for one side and black on the other side
  • Includes a cleaning roller for dust-free printing
Print 250 cards, full colour on one side with black on the reverse side.
Información de suministro: Includes 1 dye film and 1 cleaning roller.