RD90 Respirator filters for X-plore® masks
Proveedor: Dräger
6736705 6738870 6738865
DRAE6736705EA 45.2 EUR
DRAE6736705 DRAE6738870 DRAE6738865
RD90 Respirator filters for X-plore® masks
Respiradores Mascarillas reutilizables Filtros de máscaras
The filters of the Dräger X-plore® Rd90 series protect from hazardous gases and vapors, have a long life and can be used for many different masks.
  • Filters have been designed for use in the Dräger half masks, X-plore® 4390 and 4790. With the 40/90 filter box adapter they can also be used for any mask with a Rd40 connection in accordance with EN 148-1.
  • Gas filters and combination filters have a shelf-life of 6 years and, the particle filters a shelf life of 12 years from date of manufacture
  • Filters are packed individually to ensure optimum protection of the