Masterflex® Barbed High Flow+ Female Quick-Disconnect Fittings, Avantor®
Proveedor: Avantor Fluid Handling
MFLX40250-07EA 475 EUR
MFLX40250-07 MFLX40250-22 MFLX40250-37 MFLX40250-52
Masterflex® Barbed High Flow+ Female Quick-Disconnect Fittings, Avantor®
Racores y conectores
Mechanical innovations reduce risk of fluid cross-over and contamination. A mating connection is made with a High+ Flow Male fitting.
  • Aseptic connect and disconnect
  • Spring-less fluid path
  • Recessed mating features
  • USP Class VI Polypropylene and PVDF housing materials
  • Elimination of accidental disconnect with safety slide release
Masterflex® quick-disconnect fittings are an innovative and aseptic connection that allows a larger volume of fluid to pass through at lower line pressures in a smaller body. As the overall quick disconnect design has matured,More Product Information