Cylinder holder, wall mounted
Proveedor: Restek
21333
REST21333EA 115 EUR
REST21333
Cylinder holder, wall mounted
Colgadores de pared
This holder is designed to prevent free-standing gas cylinders from tipping over and injuring personnel.
  • Cast aluminum holder can be secured to a wall or the side of a work bench
  • Each mount will secure a cylinder 101,6 to 381 mm in diameter