Scandium(III) sulphate hydrate ≥99.9%
Proveedor: Thermo Fisher Scientific
42877.03
42877.06
42877.03EA
185
EUR
42877.03
42877.06
Scandium(III) sulphate hydrate ≥99.9%
Scandium(III) sulphate hydrate
Purity: 99.9%
Molecular Formula: Sc2(SO4)3.XH2O
Formula Weight: 378.10 (anhy)
Physical Form: Crystalline
Sensitivity: Hygroscopic
Appearance: White
Solubility: Freely soluble in water
MDL No.: MFCD00149854
Formula:
Sc₂(SO₄)₃·xH₂O Temperatura de almacenaje: Ambiente |
Número MDL:
MFCD00149854 Núm. CAS: 52788-54-2 |
Learn more

About VWR
Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...