Methyltriphenylsilane ≥97.0% (por GC)

Proveedor: TCI

M3138-5G M3138-25G
TCIAM3138-5GEA 52 EUR
TCIAM3138-5G TCIAM3138-25G
Methyltriphenylsilane ≥97.0% (por GC)
Methyltriphenylsilane
Formula: C₁₉H₁₈Si
Pto. Ebullición: 200°C
Número MDL: MFCD00041325
Núm. CAS: 791-29-7

Order Now

Resultados test especificaciones

Appearance White to Almost white powder to crystal
Purity (GC) min. 97.0 %
Melting point 65.0 to 69.0 deg-C


Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Más About VWR