Bel-Art Flowmi
Click to play Flowmi video See Flowmi in action
Flow Cytometry Preparation
Provides fast, efficient filtering of small samples with minimal sample loss while providing reduced risk of cytometer clogs.
Key Features and Benefits:
  • Flowmi is recommended for use with samples up to 1000µl/maximum concentration of 2MM cells/ml
  • Compact size - Highly effective with small sample volumes
  • 40µm porosity
  • Fits a wide variety of 1000µl Pipette tips
  • Sterile tips are oriented for fast attachment in space-saving tray; One-handed opening / closing