Fmoc-Dpr(Boc-Aoa)-OH ≥96.0% (by HPLC) (99.5% ee), Novabiochem®

Proveedor: Novabiochem (Part of Merck)

8.52216.0001 8.52216.0005 8.52216.0025
04-12-1185-1EA 174 EUR
04-12-1185-1 04-12-1185-5 04-12-1185-25
Fmoc-Dpr(Boc-Aoa)-OH ≥96.0% (by HPLC) (99.5% ee), Novabiochem®
Fmoc-Dpr(Boc-Aoa)-OH

An excellent derivative for the chemoselective ligation of unprotected peptides in aqueous media via oxime formation. This derivative introduces a hydroxylamine functionality which can couple with aldehyde groups present in another peptide unit.

Formula: C₂₅H₂₉N₃O₈
Núm. CAS: 600153-12-6

Order Now

Resultados test especificaciones

Alternate Names N-α-Fmoc-N-β-(N-t.-Boc-amino-oxyacetyl)-L-diaminopropionic acid
Colour (Visual) White to Slight Yellow to Beige
Appearance of Substance (Visual) Powder
Identity (IR) Passes Test
Enantiomeric Purity ≥ 99,5% (a/a)
Purity (TLC(CMA2)) ≥ 98%
Assay (HPLC, area%) ≥ 96,0% (a/a)
Solubility (1 mmole in 2 ml DMF) clearly soluble
Assay (Acidimetric) ≥ 95,0%
Water (K. F.) ≤ 2,00%
Storage Store below +30 °C


Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Más About VWR