Transition metal mix 1 for ICP, 10 elements, TraceCERT®, Supelco®
Proveedor: Merck
04330-100ML
04330-100MLEA 185 EUR
04330-100ML
Transition metal mix 1 for ICP, 10 elements, TraceCERT®, Supelco®
Normativas Estándares catiónicos Estándares de multielementos
100 mg/l each: V, Cr, Mn, Fe, Co, Ni, Cu, Zn, Ag and Cd in 2% nitric acid
Certificaciones: This certified reference material (CRM) is produced and certified in accordance with ISO/IEC 17025 and ISO 17034