di-Amonio hidrogenofosfato ≥98.0% (by acidimetry), BAKER ANALYZED® ACS, J.T.Baker®

Proveedor: Avantor
Atención

0784-01EA 0 EUR
0784-01 0784-07 0784-R
di-Amonio hidrogenofosfato ≥98.0% (by acidimetry), BAKER ANALYZED® ACS, J.T.Baker®
di-Amonio hidrogenofosfato

Crystals. Lot analysis on label.

Formula: (NH₄)₂HPO₄
Pto. de fusión: 185 °C
Densidad: 1,62 g/cm³ (20 °C)
Temperatura de almacenaje: Ambiente
Número MDL: MFCD00010891
Núm. CAS: 7783-28-0
EINECS: 231-987-8
IR: 01-2119490974-22
Índice Merck: 13,00546

Order Now

Resultados test especificaciones

Meets ACS Reagent Chemical Requirements ≥98.0%
For Laboratory,Research,or Manufacturing Use 7.7 - 8.1
Assay ((NH₄)₂HPO₄) (by acidimetry) ≥98.0 %
pH of 5% Solution at 25°C 7.7 - 8.1
Insoluble Matter ≤0.005 %
Chloride (Cl) ≤0.001 %
Nitrate (NO₃) ≤0.003 %
Sulfate (SO₄) ≤0.01 %
Heavy Metals (as Pb) ≤0.001 %
Trace Impurities - Iron (Fe) ≤0.001 %
Calcium (Ca) ≤0.001 %
Magnesium (Mg) ≤0.0005 %
Potassium (K) ≤0.005 %
Sodium (Na) ≤0.005 %
Meets Reagent Specifications for testing USP/NF monographs


Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Más About VWR