Control slides, ISOSLIDE®
Proveedor: Merck
1.00408.0001 1.00425.0001 1.02560.0001 1.00380.0001
1.00408.0001EA 301 EUR
1.00408.0001 1.00425.0001 1.02560.0001 1.00380.0001
Control slides, ISOSLIDE®
Portaobjetos Microscopy Control Slides
ISOSLIDE® control slides are designed for use with new as well as existing products.
  • Long shelf life
  • ISOSLIDE® control slides are prepared exclusively from animal material
  • All slides in a box are from the same batch and are prepared from one block
ISOSLIDE® control slides help optimise and standardise staining results by allowing typical control material to be compared with a laboratory’s own specimens. Typical material is prepared and sliced using standard procedures. One slide is already stained by the Merck staining procedure, while the other 24More Product Information