Eosina-azul de metileno según May-Grünwald modified solution stain for hematology, for cytology, Sigma-Aldrich®

Proveedor: Merck
Peligro


M-Clarity™ quality level = MQ300

1.01424.0100 1.01424.0500 1.01424.2500 1.01424.9025 1.01424.1000
1.01424.0100EA 27.8 EUR
1.01424.0100 1.01424.0500 1.01424.2500 1.01424.9025 1.01424.1000
Eosina-azul de metileno según May-Grünwald modified solution stain for hematology, for cytology, Sigma-Aldrich®
Eosina-azul de metileno según May-Grünwald

Used to stain blood, bone marrow smears, and clinical cytological specimens

Densidad: 0,79 g/cm³ (20 °C)
Número MDL: MFCD00131580
UN: 1230
ADR: 3 (6.1),II
IVDLogo

Order Now

Resultados test especificaciones

Suitability for microscopy (Blood smear) passes test
Erythrocytes pink to brownish
Nuclei violet
Eosinophilic granules red-brown
Neutrophilic granules light violet
Lymphocyte cytoplasm Blue


Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Más About VWR