Absorbente gránulos, Chemizorb® Granules
Proveedor: Merck
1.01568.1000 1.01568.5000 1.01568.9021
1.01568.1000EA 38.32 EUR
1.01568.1000 1.01568.5000 1.01568.9021
Absorbente gránulos, Chemizorb® Granules
Materiales para contención de derrames
Chemizorb granules is a coarse grained material based on chemically-inert porous minerals or synthetic co-polymers (grain size over 1 mm (95%), bulk density of 400 kg/m³).
  • Able to absorb up to 100% of its own weight
  • Low viscosity liquids are absorbed instantaneously, highly viscous media (glycerine, oils) within a few minutes
  • Good dosing is possible thanks to the coarse grained format
  • Granule form is used in all those places where it is not suitable to use the powder form, for example, in draughty rooms or outside
Es insoluble en agua y en todos losMore Product Information
Limited Time Sale
Select items below are currently on sale