TLC plates, kieselguhr and mixed layers
Proveedor: Merck
1.05567.0001 1.05568.0001 1.05738.0001
1.05567.0001EA 233 EUR
1.05567.0001 1.05568.0001 1.05738.0001
TLC plates, kieselguhr and mixed layers
Placas TLC
Kieselguhr is a natural diatomaceous earth that can be used for the separation of polar or moderately polar compounds. For the mixed layer plates a combination of classical silica gel 60 and kieselguhr is used, providing favorable properties for certain applications such as separations of inorganic ions, herbicides or steroids.