Ácido trifluoroacético, Uvasol® para espectroscopia, Supelco®

Proveedor: Merck
Peligro


M-Clarity™ quality level = MQ100

1.08262.0025 1.08262.0100 1.08262.1000 1.08262.2500
1.08262.0025EA 51.6 EUR
1.08262.0025 1.08262.0100 1.08262.1000 1.08262.2500
Ácido trifluoroacético, Uvasol® para espectroscopia, Supelco®
Ácido trifluoroacético

Accurate analytic results in UV/Vis and infrared spectroscopy depend on the use of very pure solvents for sample preparation. The Uvasol® solvents range has been specially designed for spectroscopy and other applications requiring solvents of the highest spectral purity. The refinement process allows a greater degree of security in applications and avoids misinterpretation of analytical results caused by traces of UV, IR and fluorescence contamination. Uvasol® solvents offer best UV transmittance. In all specifications the minimum transmittance for five typical wavelengths are identified. Furthermore the transmittance is specified in accordance with Reag. Ph. Eur. and ACS.

Formula: C₂HF₃O₂
Pto. Ebullición: 71,8 °C (1013 hPa)
Pto. de fusión: –15 °C
Densidad: 1,490 g/cm³ (20 °C)
Temperatura de almacenaje: Ambiente
Número MDL: MFCD00004169
Núm. CAS: 76-05-1
EINECS: 200-929-3
UN: 2699
ADR: 8,I
Índice Merck: 13,09754
https://digitalassets.avantorsciences.com/adaptivemedia/rendition?id=9501f3ac85d66e8d19d1cd1c3fe5409b974e9d8a&vid=be6aa627129b49ccda76006e7003b46d79b933cc&prid=web&clid=SAPDAM

Order Now

Resultados test especificaciones

Solubility (20 °C) soluble
Melting Point -15 °C
Molar Mass 114.02 g/mol
Boiling Point 72 °C (1013 hPa)
Vapor Pressure 141 hPa (20 °C)
Density 1.48 g/cm³ (20 °C)
pH 1 (10 g/L, H₂O)
Hazard Class 8
Hazard Class Description Corrosive material


Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Más About VWR