Ácido clorhídrico 0.5 mol/l (0.5 N), Titripur® Reag. USP solución volumétrica, Supelco®

Proveedor: Merck
Atención

Sinónimos: Hydrochloric acid solution, Muriatic acid

M-Clarity™ quality level = MQ100

1.09058.1000 1.09058.9025 1.09058.5000 1.09058.4000 1.09058.1003
1.09058.1000EA 17.71 EUR
1.09058.1000 1.09058.9025 1.09058.5000 1.09058.4000 1.09058.1003
Ácido clorhídrico 0.5 mol/l (0.5 N), Titripur® Reag. USP solución volumétrica, Supelco®
Ácido clorhídrico

Merck have introduced into the range of Titripur® Titrants some packs with additional technology - Supelco® SmartTitrants that use an RFID tag to provide a seamless data transfer between the smart titrator and the RFID tag on the label of the bottle of titrant. It will ensure that all information needed for method execution, result calculation and documentation is transferred to the titrator equipped with this technology. MettlerToledo Compact and Excellence titrators include this technology. Please see ordering table below for packs with RFID tag.


Stability of product in Titripac packaging: Merck have carried out testing to show that products are stable for 36 months when in use and opened. Please see Technical report (Product specification) in the Related Information tab below.

Formula: HCl
Temperatura de almacenaje: Ambiente
Número MDL: MFCD00011324
Núm. CAS: 7647-01-0
EINECS: 231-595-7
UN: 1789
ADR: 8,II

Order Now


Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Más About VWR