Bario perclorato 0.005 mol/l in 2-propanol/water (80/20), Titripur® Reag. USP solución volumétrica, Supelco®

Proveedor: Merck
Peligro


M-Clarity™ quality level = MQ100

1.09086.1000 1.09086.1003
1.09086.1000EA 93.6 EUR
1.09086.1000 1.09086.1003
Bario perclorato 0.005 mol/l in 2-propanol/water (80/20), Titripur® Reag. USP solución volumétrica, Supelco®
Bario perclorato

Merck have introduced into the range of Titripur® Titrants some packs with additional technology - Supelco® SmartTitrants that use an RFID tag to provide a seamless data transfer between the smart titrator and the RFID tag on the label of the bottle of titrant. It will ensure that all information needed for method execution, result calculation and documentation is transferred to the titrator equipped with this technology. MettlerToledo Compact and Excellence titrators include this technology. Please see ordering table below for packs with RFID tag.

Formula: Ba(ClO₄)₂
Número MDL: MFCD00003440
Núm. CAS: 13465-95-7
UN: 1993
ADR: 3,II

Order Now


Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Más About VWR