Potasio hidróxido 0.1 mol/l (0.1 N) in ethanol, Titripur® Reag. Ph. Eur., Reag. USP solución volumétrica, Supelco®

Proveedor: Merck
Peligro


M-Clarity™ quality level = MQ100

1.09115.1000 1.09115.2500 1.09115.1000_T 1.09115.2500_T 1.09115.1003
1.09115.1000EA 79.3 EUR
1.09115.1000 1.09115.2500 MERC1.09115.1000_T MERC1.09115.2500_T 1.09115.1003
Potasio hidróxido 0.1 mol/l (0.1 N) in ethanol, Titripur® Reag. Ph. Eur., Reag. USP solución volumétrica, Supelco®
Potasio hidróxido

Merck have introduced into the range of Titripur® Titrants some packs with additional technology - Supelco® SmartTitrants that use an RFID tag to provide a seamless data transfer between the smart titrator and the RFID tag on the label of the bottle of titrant. It will ensure that all information needed for method execution, result calculation and documentation is transferred to the titrator equipped with this technology. MettlerToledo Compact and Excellence titrators include this technology. Please see ordering table below for packs with RFID tag.

Formula: KOH
Densidad: 0,836 g/cm³ (20 °C)
Pto. de inflamación: 12 °C (EtOH)
Temperatura de almacenaje: Ambiente
Número MDL: MFCD00003553
Núm. CAS: 1310-58-3
EINECS: 215-181-3
UN: 2924
ADR: 3 (8),II

Order Now

Resultados test especificaciones

Form liquid
Amount-of-substance concentration 0.0995-0.1005
Measurement uncertainty +/- 0.0003
Traceability NIST SRM

ESPECIFICACIONES

Environmentally Preferable

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Más About VWR