Potasio dicromato 1/24 mol/l (0.25 N) en solución acuosa, Supelco®

Proveedor: Merck
Peligro


M-Clarity™ quality level = MQ100

1.09118.1000 1.09118.1003
1.09118.1000EA 47.2 EUR
1.09118.1000 1.09118.1003
Potasio dicromato 1/24 mol/l (0.25 N) en solución acuosa, Supelco®
Potasio dicromato

Merck have introduced into the range of Titripur® Titrants some packs with additional technology - Supelco® SmartTitrants that use an RFID tag to provide a seamless data transfer between the smart titrator and the RFID tag on the label of the bottle of titrant. It will ensure that all information needed for method execution, result calculation and documentation is transferred to the titrator equipped with this technology. MettlerToledo Compact and Excellence titrators include this technology. Please see ordering table below for packs with RFID tag.

Formula: K₂Cr₂O₇
MW: 294.18 g/mol
Pto. Ebullición: 500
Pto. de fusión: 398
Temperatura de almacenaje: Ambiente
Número MDL: MFCD00011367
Núm. CAS: 7778-50-9
EINECS: 231-906-6
UN: 3287
Índice Merck: 12,07790

Order Now

Resultados test especificaciones

Solubility (20 °C) soluble
Density 1.01 g/cm3 (20 °C)
pH 3.8 (H2O, 20 °C)


Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Más About VWR