Etanol ≥99.9% (por GC), LiChrosolv®, grado gradiente for liquid chromatography, Supelco®

Proveedor: Merck
Peligro


M-Clarity™ quality level = MQ100

1.11727.1000 1.11727.2500 1.11727.4000 1.11727.9030 1.11727.9185 1.11727.1000_T 1.11727.2500_T 1.11727.4000_T 1.11727.9030_T 1.11727.0500
1.11727.1000EA 159 EUR
1.11727.1000 1.11727.2500 1.11727.4000 1.11727.9030 1.11727.9185 MERC1.11727.1000_T MERC1.11727.2500_T MERC1.11727.4000_T MERC1.11727.9030_T 1.11727.0500
Etanol ≥99.9% (por GC), LiChrosolv®, grado gradiente for liquid chromatography, Supelco®
Etanol

With their high degree of UV transmittance, low particle count, low acidity and alkalinity and low evaporation residue level, LiChrosolv® solvents are ideal for reproducible separations. Since separations are normally carried out under gradient conditions in analytical HPLC, we offer solvents in 'gradient grade' as well as 'isocratic grade'. This minimises the gradient effect of the solvent involved.

Formula: C₂H₅OH
Pto. Ebullición: 78,3 °C (1013 hPa)
Pto. de fusión: –117 °C
Densidad: 0,7895 g/cm³ (20 °C)
Pto. de inflamación: 12 °C
Temperatura de almacenaje: Ambiente
Número MDL: MFCD00003568
Núm. CAS: 64-17-5
EINECS: 200-578-6
UN: 1170
ADR: 3,
IR: 01-2119457610-43
Índice Merck: 12,03806

Order Now

Resultados test especificaciones

Purity (GC) ≥ 99.9 %
Identity (IR) conforms
Evaporation residue ≤ 2.0 mg/l
Water ≤ 0.1 %
Acidity ≤ 0.0002 meq/g
Alkalinity ≤ 0.0002 meq/g
Gradient grade (at 235 nm) ≤ 5 mAU
Gradient grade (at 254 nm) ≤ 2 mAU
Transmission (at 225 nm) ≥ 60 %
Transmission (at 240 nm) ≥ 85 %
Transmission (from 260 nm) ≥ 98 %
Filtered by 0.2 µm filter
Synonyms Ethyl alcohol, EtOH
Hill Formula C2H6O
Chemical Formula C2H5OH


Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Más About VWR