IR3535® (Insect repellent 3535)

Proveedor: Merck
Atención

Sinónimos: N-Acetil-N-butil-β-alaninato de etilo

1.11887.0100 1.11887.1000 1.11887.9025 1.11887.9910
1.11887.0100EA 0 EUR
1.11887.0100 1.11887.1000 1.11887.9025 1.11887.9910
IR3535® (Insect repellent 3535)
Ethyl butylacetylaminopropionate

IR3535® (Insect repellent 3535) 3-[N-n-butyl-N-acetyl] aminopropionic acid ethylester


  • Insect repellent biocidal active substance
  • Application Cosmetics

Formula: C₁₁H₂₁NO₃
MW: 215.29 g/mol
Pto. Ebullición: 292
Densidad: 0.998 g/cm³ (20 °C)
Pto. de inflamación: 159
Temperatura de almacenaje: Ambiente
Número MDL: MFCD00214076
Núm. CAS: 52304-36-6
EINECS: 257-835-0

Order Now

Resultados test especificaciones

Appearance Clear, colorless to slightly yellowish liquid; almost odorless
Identification (IR) IR-Spectrum Conform;
Assay Not less than 99.0 %
Appearance of sol (5%, water) Clear and colorless
pH (5%, water) 4.0-6.0
Free acid (as acetic acid) Not more than 0.1 %
Water (K.F.) Not more than 0.5 %
Refractive index 1.452-1.455
Sulphated ash Not more than 0.1 %
Total viable aerobic count Not more than 100 CFU/g
Escherichia coli Absence in 1 g
Pseudomonas aeruginos Absence in 1 g
Staphylococcus aureus Absence in 1 g
Candida albicans Absence in 1 g
Salmonella Absence in 10 g
Microbiological tests periodically performed


Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Más About VWR