Acetonitrilo, Prepsolv® for preparative chromatography

Proveedor: Merck
Peligro

Sinónimos: Cianuro de metilo

M-Clarity™ quality level = MQ200

1.13358.2500 1.13358.9030 1.13358.9185 1.13358.9910 1.13358.9186 1.13358.9913 1.13358.2509
1.13358.2500EA 275 EUR
1.13358.2500 1.13358.9030 1.13358.9185 1.13358.9910 1.13358.9186 1.13358.9913 1.13358.2509
Acetonitrilo, Prepsolv® for preparative chromatography
Acetonitrilo

Prepsolv® solvents are tailored to the requirements of preparative HPLC to facilitate scale-up from analytical to preparative separations. With their extremely low evaporation residue (< 1 mg/l) and low water content, they ensure optimal protection for columns.

Formula: H₃CCN
MW: 41.05 g/mol
Pto. Ebullición: 81.6 °C (1013 hPa)
Pto. de fusión: –45.7 °C
Densidad: 0.782 g/cm³ (20 °C)
Pto. de inflamación: 2 °C (copa cerrada)
Temperatura de almacenaje: Ambiente
Número MDL: MFCD00001878
Núm. CAS: 75-05-8
EINECS: 200-835-2
UN: 1648
ADR: 3
IR: 01-2119471307-38
Índice Merck: 13,00071
https://digitalassets.avantorsciences.com/adaptivemedia/rendition?id=cef8be50fcca82050c5bda827ad9c5f57f4c5e27&vid=b80b5c39e654fcf6904b18b58e22a816e8e138b7&prid=web&clid=SAPDAM

Order Now


Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Más About VWR