Acetonitrilo, LiChrosolv®, grado isocrático for liquid chromatography, Supelco®

Proveedor: Merck
Peligro


M-Clarity™ quality level = MQ100

1.14291.1000 1.14291.2500 1.14291.4000 1.14291.9010 1.14291.9030 1.14291.9185 1.14291.9910
1.14291.1000EA 139 EUR
1.14291.1000 1.14291.2500 1.14291.4000 1.14291.9010 1.14291.9030 1.14291.9185 1.14291.9910
Acetonitrilo, LiChrosolv®, grado isocrático for liquid chromatography, Supelco®
Acetonitrilo

With their high degree of UV transmittance, low particle count, low acidity and alkalinity and low evaporation residue level, LiChrosolv® solvents are ideal for reproducible separations. Since separations are normally carried out under gradient conditions in analytical HPLC, we offer solvents in "gradient grade" as well as "isocratic grade". This enables to minimize the gradient effect of the solvent involved.

Formula: H₃CCN
Pto. Ebullición: 81,6 °C (1013 hPa)
Pto. de fusión: –45,7 °C
Densidad: 0,782 g/cm³ (20 °C)
Pto. de inflamación: 2 °C (copa cerrada)
Temperatura de almacenaje: Ambiente
Número MDL: MFCD00001878
Núm. CAS: 75-05-8
EINECS: 200-835-2
UN: 1648
ADR: 3,
IR: 01-2119471307-38
Índice Merck: 13,00071
https://digitalassets.avantorsciences.com/adaptivemedia/rendition?id=cef8be50fcca82050c5bda827ad9c5f57f4c5e27&vid=b80b5c39e654fcf6904b18b58e22a816e8e138b7&prid=web&clid=SAPDAM

Order Now


Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Más About VWR