Sodio hidróxido 0.1 mol/L en solución acuosa, EMPROVE® EXPERT para producción biofarmacéutica

Proveedor: MERCK PRODUCTION CHEMICALS

Sinónimos: Caustic soda

Filtrado por 0,2 µm

1.37058.9950 1.37058.9025 1.37058.0001
1.37058.9950EA 0 EUR
1.37058.9950 1.37058.9025 1.37058.0001
Sodio hidróxido 0.1 mol/L en solución acuosa, EMPROVE® EXPERT para producción biofarmacéutica
Sodio hidróxido

This product has been manufactured from Sodium hydroxide pellets Ph Eur, BP, JP, NF and waterfor injection (wfi) ace. USP which conforms with highly purified water (hpw) ace. Ph Eur.


Elemental impurity specifications have been set considering ICH Q3D (Guideline for Elementallmpurities). Class 1-3 elements are not likely to be present above the ICH Q3D option 1 limit, unless specified and indicated (*).

Formula: NaOH
Densidad: 1 g/cm³ (25 °C)
Temperatura de almacenaje: Ambiente
Número MDL: MFCD00003548
Núm. CAS: 1310-73-2
EINECS: 215-185-5
UN: 1824
ADR: 8,

Order Now

Resultados test especificaciones

Assay (acidimetric,NaOH) 0.05- 0.15 mol/l
Carbonate (as Na2CO3) ≤ 0.2%
Chloride (Cl) ≤ 5%
Sulfate (SO.) ≤ 30 ppm
Heavy metals (as Pb) ≤ 5.0 ppm
Al (Aluminium) ≤ 10 ppm
Fe (Iron) ≤ 3 ppm
Residual solvents (ICH Q3C) excluded by production process
Endotoxins ≤ 2.0 I.U./ml


Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Más About VWR