Bovine serum albumin (BSA)

Proveedor: SEQENS

1005-30-100ML 1005-30-500ML 1005-30-1L 1005-30-5L
1005-30-100MLEA 61.3 EUR
1005-30-100ML 1005-30-500ML 1005-30-1L 1005-30-5L
Bovine serum albumin (BSA)
Proteínas y Péptidos

SEQENS IVD manufactures Bovine Serum Albumin (BSA) derived from plasma, collected from healthy animals of French origin. This product is obtained by 'Heat Shock' method, which ensures higher purity and preserves BSA intrinsic properties.


  • Albumin purity: >97%
  • Total proteins: >290 g/L
  • Proteases: Undetectable
  • Shelf life: 3 years
  • Sterile filtered 0,2 µm


Full traceability and batch-to-batch repeatability guaranteed.


Información de suministro: Supplied as a ready to use 30% solution in deionised water.

Order Now

ESPECIFICACIONES

Nombre de proteína / péptido Bovine Serum Albumin
Environmentally Preferable
Sinónimos de proteínas BSA|Fraction V|Cohn Fraction V
Especies Bovine
Fuente Serum
Sin proteasa Y
Concentración 30% solution

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Más About VWR