Blotting membranes, Amersham™ Hybond™ 0.2 PVDF
Proveedor: Cytiva
10600058 10600021 10600057 10600101 10600122
10600058.EA 323 EUR
10600058. 10600021. 10600057. 10600101. 10600122.
Blotting membranes, Amersham™ Hybond™ 0.2 PVDF
Membranas de transferencia
Amersham™ Hybond™ 0.2 PVDF is a 0,2 μm pore size hydrophobic membrane with high physical strength. This results in significant handling advantages over unsupported nitrocellulose and makes the membrane highly suitable for stripping and reprobing. The small pore size of the membrane minimises 'blow-through' and increases protein binding over a wide range of molecular weights. Ideal for use with standard colorimetric and chemiluminescent detection methods for proteins of
  • High protein retention, even after harsh washing steps
  • Protein binding capacity over 200