Powered air-purifying respirator (PAPR), X-plore 8000
Proveedor: Dräger
R59632 R59640 R59650 R59670 R59500 R59631 R59600 R59610 R59620 R59630 R59530 R59563 R59564 R59565 R59566 R59585 R59690 R59700 R59710 R59730 R59750 R59760 R59800 R59830 R59820 R59826 R59850 R59810 R59840 R59860 R59862 R59863 R59901 R59902 R59903 R59904 R59870 3710775 3710774 3710800 3710805 R59545 3710785 R59560 3710776 R59950 R59890 3710779 R59540
111-1812EA 15.3 EUR
111-1812 111-1813 111-1814 111-1815 111-1799 111-1811 111-1807 111-1808 111-1809 111-1810 111-1800 111-1801 111-1802 111-1803 111-1804 111-1806 111-1816 111-1817 111-1818 111-1819 111-1820 111-1821 111-1822 111-1826 111-1824 111-1825 111-1828 111-1823 111-1827 111-1829 111-1854 111-1853 111-1856 111-1857 111-1858 111-1855 111-1850 DRAE3710775 DRAE3710774 DRAE3710800 DRAE3710805 DRAER59545 DRAE3710785 DRAER59560 DRAE3710776 DRAER59950 DRAER59890 DRAE3710779 DRAER59540
Powered air-purifying respirator (PAPR), X-plore 8000
Respiradores Sistemas de respiración
The powered air-purifying respirator (PAPR) X-plore 8000 offers a number of user friendly standard and specialised carrying systems for standard and decontamination applications. Dräger has also developed a wide range of tight- and loose-fitting headgear such as half and full-face masks, short and long hoods and helmets with visor and protective visor. Standard and enhanced flexibility hoses are available and compatibility is ensured. Modular components: A wide range of components for every application.
  • Heavy-duty design with rubber protectors and high ingress protection (IP 65)