Mascarillas autofiltrantes para partículas, desechable, FFP1/FFP2/FFP3
Proveedor: 3M
9914 9926 9928 9913 9936 9922
111-8935EA 115 EUR
111-8935 111-8938 111-0030 111-0031 111-0452 111-0917
Mascarillas autofiltrantes para partículas, desechable, FFP1/FFP2/FFP3
Respiradores Mascarillas desechables Mascarillas para partículas
This range provides lightweight, effective, comfortable and hygienic protection from dust and aerosols.
  • Convex design, double strap, foam nose protector and aluminium nose piece for secure fitting on any face shape
  • Patented 3M™ Cool Flow™ valve on models 'with valve' and collapse resistant shell available on all models
  • No expensive or time-intensive maintenance is required
The masks also protect against low concentrations (nuisance levels) of organic fumes or acid gases such as sulphur dioxide or hydrogen fluoride, below the Workplace Exposure Limit (More Product Information