Nitrato de itrio (III) hidrato ≥99.99% (REO, rare earth oxide basis)

Proveedor: Thermo Fisher Scientific
Peligro

Sinónimos: Yttrium trinitrate hydrate

11187.06 11187.14 11187.22 11187.59
11187.06EA 47.2 EUR
11187.06 11187.14 11187.22 11187.59
Nitrato de itrio (III) hidrato ≥99.99% (REO, rare earth oxide basis)
Nitrato de itrio (III) hidrato

Purity: 99.99% Molecular Formula: Y(NO3)3.xH2O Formula Weight: 274.92(anhy) Physical Form: Crystalline Aggregates Sensitivity: Hygroscopic Merck Reference: 14,10107 MDL No.: MFCD00011469

Formula: N₃O₉Y·H₂O
MW: 274.92 g/mol (anhy)
Temperatura de almacenaje: Ambiente
Número MDL: MFCD00011469
Núm. CAS: 140191-40-8
EINECS: 233-802-6
UN: 1477
ADR: 5.1 III

Order Now


Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Más About VWR