Guantes para salas blancas resistentes al ácido, de neopreno/nitrilo, Advantech 517
Proveedor: MAPA
34517038 34517036 34517037 34517039 34517030
112-0503EA 66 EUR
112-0503 112-0501 112-0502 112-0504 112-0500
Guantes para salas blancas resistentes al ácido, de neopreno/nitrilo, Advantech 517
Guantes
These powder-free cleanroom gloves are made from Tri-polymer blend (nitrile, neoprene, and natural rubber) which offers excellent resistance and strong chemical protection against a great number of chemical products. Free from silicone or paraffin.
  • Extreme comfort and durability, exceptional chemical and mechanical resistance
  • Resistant to etchants, acids and caustics
  • Embossed texture for good grip of wet and dry objects
  • Wide cuffs enable double-gloving
Suitable for use in Class 100, ISO 5 or M3.5 cleanrooms.
Certificaciones: EN 388:2016 (1110X), ENMore Product Information