Guantes para sala limpia, policloropreno, estériles, BioClean Ultimate™
Proveedor: Nitritex
BUPS-80 BUPS-75 BUPS-90 BUPS-60 BUPS-65 BUPS-70 BUPS-85
112-1664EA 620 EUR
112-1664 112-1663 112-1666 112-1660 112-1661 112-1662 112-1665
Guantes para sala limpia, policloropreno, estériles, BioClean Ultimate™
Guantes
BioClean Ultimate™ sterile cleanroom gloves are manufactured from polychloroprene and are hand-specific with a beaded cuff. They offer excellent cytotoxic protection with sensitivity.
  • Sterile, powder-free, textured finish provides a good grip
  • With no latex proteins, Type 1 latex allergic reactions are eliminated
  • Anatomical hand-specific shape for high tactility, protection and comfort
  • Fit and feel of natural latex gloves
Adecuado para su uso en salas limpias de Clase 10, ISO 4 o M2.5 y EU GMP Grado A.
EN 420, 455, Cat III, AQL 1.5