Cut resistant gloves, Krytech 586
Proveedor: MAPA
34586036 34586030 34586038 34586031 34586039 34586037
112-2815EA 18.3 EUR
112-2815 112-2813 112-2817 112-2814 112-2818 112-2816
Cut resistant gloves, Krytech 586
Guantes
This high performance glove, made of PU with a seamless textile support from HDPE fibres, provides protection for intricate handling in reasonably clean environments.
  • HDPE fibres ensure excellent cut resistance (level 5) with reduced thickness for good dexterity
  • Ventilated back for breathability
  • Visible indicator in blue textile to easily identify the cut-performance level in the workplace
Suitable when handling sharp objects and large metal plates, for example in the automotive or mechanical industry.
Certificaciones: EN 388 (4X43D); Cat. II
Order Now
ESPECIFICACIONES
Modelo Krytech 586
Environmentally Preferable
Ambidiestro/Específico para cada mano Específico para cada mano
Desechable / Reutilizable Reutilizable
Material del revestimiento HDPE
Tipo de puño Puño elástico
Normativa EN EN 388 (4X43D)