VWR® Cleanroom Gloves, Soft Nitrile, Non Sterile, ISO 4, 400 mm
Proveedor: VWR Collection
112-5119EA 544 EUR
112-5119 112-5122 112-5124 112-5121 112-5123 112-5120
VWR® Cleanroom Gloves, Soft Nitrile, Non Sterile, ISO 4, 400 mm
Guantes
VWR® cleanroom soft, nitrile gloves are manufactured from carboxylated acrylonitrile butadiene that provides reliable, durable and comfortable hand protection during demanding cleanroom applications.
  • Designed for elbow length protection
  • Ambidextrous, designed with micro textured fingers and palm, beaded cuff
  • Not made from natural rubber latex
  • Static-disspative
  • Single-use only
Recommended for ISO Class 4 (FED-STD-209E Class 10) or higher cleanrooms.
Certificaciones: EN ISO 374-1 Type B (KPT), EN ISO 374-5; Cat. III; AQL 1,5
ASTM D412; ASTM D257,More Product Information