Industrial gloves, Titan 375 / 376
Proveedor: MAPA
34375016 34375018 34375017 34375019
112-5566EA 84.4 EUR
112-5566 112-5568 112-5567 112-5569
Industrial gloves, Titan 375 / 376
Guantes
These nitrile gloves with textile lining and smooth finish provide protection against humidity, oil and grease splashes.
  • Good abrasion resistance
  • Exceptionally comfortable lining: no seams on the working surfaces
  • Last longer than precision work gloves, yet offers better comfort and dexterity than heavy-duty gloves
Ideal for precision mechanical work, handling oily mechanical parts, wood working, packaging and manufacturing small electrical components.
Certificaciones: EN 388:2016 (3111X), Cat. II
Order Now
ESPECIFICACIONES
Modelo Titan 375
Environmentally Preferable
Ambidiestro/Específico para cada mano Específico para cada mano
Desechable / Reutilizable Reutilizable
Material del revestimiento Algodón
Tipo de puño Pinked
Normativa EN EN 388:2016 (3111X)