General purpose gloves, Ultranitril 454
Proveedor: MAPA
34454136 34454137 34454008 34454009 34454130
112-5585EA 5.9 EUR
112-5585 112-5586 112-5587 112-5588 112-5584
General purpose gloves, Ultranitril 454
Guantes
These synthetic elastomer gloves with cotton flocked interior are designed for use in mildly aggressive environments.
  • Flocked interior provides good absorption of perspiration
  • Outstanding precision dexterity
  • Embossed texture provides good handling of slippery objects
  • Optimal comfort: anatomical shape, new flocking quality, liquidproof
Ideal for common tasks in hospitals and clinics and cleaning tasks involving household detergents.
Certificaciones: EN 388:2016 (2000X), EN ISO 374-1 Type B (KPT), EN ISO 374-5; Cat. III