General purpose gloves, Showa 310
Proveedor: Showa
310OR-10/XL 310G-6/XS 310G-7/S 310G-8/M 310OR-8/M 310G-9/L 310OR-9/L 310G-10/XL 310B-10/XL 310B-7/S 310B-8/M 310B-9/L 310OR-7/S
112-5776EA 53.1 EUR
112-5776 112-5772 112-5773 112-5774 112-5778 112-5775 112-5779 112-5771 112-5767 112-5768 112-5769 112-5770 112-5777
General purpose gloves, Showa 310
Guantes
These hand-specific cotton gloves with latex coating on palm and fingertips are flexible and robust offering great dexterity, outstanding grip and good resistance to tearing. They are ideal for heavy duty manual handling.
  • Latex coating provides protection against abrasions, alcohols and aggressive detergents
  • Excellent grip in damp and dry environments
  • Seamless knit to prevent irritation
  • Breathable back of hand to reduce perspiration
  • Designed for easy movement and continuous wear thanks to renowned ergonomic shaped hand mould
Ideal for use inMore Product Information
Order Now
ESPECIFICACIONES
Modelo 310G 310B 310OR
Environmentally Preferable
Ambidiestro/Específico para cada mano Específico para cada mano Específico para cada mano Específico para cada mano
Desechable / Reutilizable Reutilizable Reutilizable Reutilizable
Latex/No contiene Latex Latex Latex Latex
Material del revestimiento Algodón/Poliéster Algodón/Poliéster Algodón/Poliéster
Acabado Palm-dipped natural rubber coating Palm-dipped natural rubber coating Palm-dipped natural rubber coating
Calibre 10 10 10
Tipo de puño Puño elástico Puño elástico Puño elástico
Normativa EN EN 388:2016 (2142X) EN 388:2016 (2142X) EN 388:2016 (2142X)