Chemical resistant gloves, Showa 874 / 874R
Proveedor: Showa
874-07 874-08 874-09 874R-09 874R-10 874R-07 874R-08 874R-11 874-10 874-11
112-5942EA 62.4 EUR
112-5942 112-5943 112-5944 112-5948 112-5949 112-5946 112-5947 112-5950 112-5945 BEST874-11
Chemical resistant gloves, Showa 874 / 874R
Guantes
These thin, light gloves with butyl coating protect the hand from a wide array of chemical hazards. Available with a smooth finish or with a rough finish on the fingertips.
  • Impermeable for working in damp or greasy environments
  • Rolled cuff prevents dirt from entering the glove
  • Without special treatment: Minimal allergy risks
  • Designed for easy movement and continuous wear
  • Wrist well protected
Ideal for applications in automotive and chemical industries, in metallurgy and in the public sector.
Certificaciones: EN 388:2016 (0010X), EN ISO 374-1 Type BMore Product Information
Order Now
ESPECIFICACIONES
Modelo 874 874R
Environmentally Preferable
Ambidiestro/Específico para cada mano Específico para cada mano Específico para cada mano
Desechable / Reutilizable Reutilizable Reutilizable
Acabado Smooth Rough
Tipo de puño Puño con reborde Puño con reborde
Normativa EN EN 388:2016 (0010X), EN ISO 374-1 Type B (BIK), EN ISO 374-5 VIRUS EN 388:2016 (0010X), EN ISO 374-1 Type B (BIK), EN ISO 374-5 VIRUS