Heat resistant gloves, Showa 8814
Proveedor: Showa
8814/10 8814/7 8814/8 8814/9
112-5951EA 24.9 EUR
112-5951 112-5952 112-5953 112-5954
Heat resistant gloves, Showa 8814
Guantes
These non woven liners with full neoprene coating protect the hand against hot temperatures and abrasion while providing a good grip.
  • Impermeable for working in damp or greasy environments
  • Slip-on designed gauntlet: Easy to put on and remove
  • Textured finish: Surface provides better grip
  • Minimal allergy risks
  • Wrist well protected
  • Low-soil colour
Ideal for use in the automotive or food industry.
Certificaciones: EN 388:2016 (2242X), EN 407 (X2XXXX); Cat. II
Food contact approved.
Precaución: Gloves must not come in contact with an open flame.