Chemical resistant gloves, Showa KV660
Proveedor: Showa
KV660-9/L KV660-11/XXL KV660-10/XL KV660-8/M
112-5998EA 172 EUR
112-5998 112-5996 112-5995 112-5997
Chemical resistant gloves, Showa KV660
Guantes
Providing defense against a broad range of hazards these cut and chemical-resistant gloves are forged with triple-dipped PVC and a reinforced Kevlar® liner.
  • Rough surface finish offers a good grip and increased durability
  • Even after multiple washes, the KV660 maintains its barrier to hazards
  • Seamless Aramid/Kevlar® knit
The KV660 provides first-class defense against oils, chemicals, abrasion, and cuts.
Typical applications include: Chemical spray and treatment, coating preparation, painting & spray workshops, bottling, HVAC, plumbing, gutteringMore Product Information