Relleno para guantes resistentes a los cortes, Whizard™
Proveedor: WELLS LAMONT
M281L M281M M281S M281XL
112-1195EA 132 EUR
112-1195 112-1196 112-1197 112-6365
Relleno para guantes resistentes a los cortes, Whizard™
Guantes
Great for use as a cut-resistant cleanroom glove liner.
  • Highly reusable
  • Made in USA
Certificaciones: ANSI A4, level C cut resistance. Category III for particulates.