Cleanroom gloves, latex, Kimtech™ G3
Proveedor: KIMBERLY CLARK
56813 56814 56815 56816
112-6406EA 372 EUR
112-6406 112-6405 112-6404 112-6403
Cleanroom gloves, latex, Kimtech™ G3
Guantes
This high-quality ambidextrous glove with textured palm and fingertips provides improved grip and comfort in both wet / dry conditions.
  • Disposable, non sterile
  • Good grip and comfort, enhanced tactile sensitivity and performance
  • Powder-free construction minimises contamination and drying effects on the wearer’s skin, ensuring that cleanroom users are protected as much as their applications and equipment
  • Beaded cuffs for easy donning, and better seal formation with other garments
Kimtech™ G3 latex ambidextrous cleanroom gloves provide enhanced tactileMore Product Information