Cleanroom gloves, nitrile, sterile, PUREZERO® HG3
Proveedor: Halyard
CLN923210 CLN923260 CLN923265 CLN923270 CLN923275 CLN923280 CLN923285 CLN923290
112-6494EA 562 EUR
112-6494 112-6495 112-6496 112-6497 112-6498 112-6499 112-6500 112-6501
Cleanroom gloves, nitrile, sterile, PUREZERO® HG3
Guantes
High quality gloves that are designed to exact standards to meet the requirements of a contamination controlled environment.
  • High quality nitrile
  • Sterile
  • Accelerator-free formula reduces risk of type IV allergies
  • For use in cleanroom environments (ISO 4/5, grade A/B aseptic)
  • REACH Annex XVII
Offering the right glove for the right controlled environment, PUREZERO® provides a cleanroom glove with guaranteed low particulate counts and endotoxin levels due to repeated washes in deionised water and chlorination. Nickel-, powder-, silicon- and latex-free,More Product Information