Neodymium(III) chloride hydrate ≥99.9% (REO, rare earth oxide basis), REacton®

Proveedor: Thermo Fisher Scientific
Atención

Sinónimos: Neodymium trichloride hydrate

11251.18 11251.30
11251.18EA 127 EUR
11251.18 11251.30
Neodymium(III) chloride hydrate ≥99.9% (REO, rare earth oxide basis), REacton®
Neodymium(III) chloride hydrate

Neodymium(III) chloride hydrate is used for production of neodymium metal. It also finds application in lasers and fibre amplifiers. It serves as dopants for silica based as well as plastic fibres. It plays an important role of additives in organic light emitting diodes resulting in improved LED emission spectrum. In addition, it is used in catalysis of polymerisation of various dienes. It is used to modify titanium dioxide for use as a photocatalyst. Its coating on aluminium and its alloy protect them from corrosion. It is also extensively used as fluorescent labels for organic molecules.


Purity: 99.9% CAS No.: 10024-93-8 Molecular Formula: NdCl3.xH2O Formula Weight: 250.60(anhy) Physical Form: Crystalline Aggregates Odour: Odourless Merck Reference: 14,6450 MDL No.: MFCD00011132

Formula: NdCl₃·xH₂O
MW: 250.60 g/mol (anhy)
Pto. de fusión: 124 °C
Densidad: 2.28 g/cm³ (20 °C)
Temperatura de almacenaje: Ambiente
Número MDL: MFCD00011132
Núm. CAS: 13477-89-9
EINECS: 233-031-5
Índice Merck: 14,6450

Order Now


Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Más About VWR