VWR® Basic, Bouffant Caps, Latex-Free
Proveedor: VWR Collection
113-1459EA 69.3 EUR
113-1459 113-1460 113-8762 113-8877 113-8887 113-8886 113-8888 113-8874 113-8875 113-8876 113-8879 113-8882 113-8880 113-8881 113-8883 113-8878 113-8884 113-8885
VWR® Basic, Bouffant Caps, Latex-Free
Prendas desechables
These disposable caps are made from a non woven, spunbonded polypropylene (SPP) fabric, which protects against dirt, grime, and certain dry particulates, in non hazardous environments.
  • Free from natural rubber latex
  • Available in a wide range of colours
Weight: 10 g/m²
Envase: 100 pieces per bag, 10 bags per case.
Order Now
ESPECIFICACIONES
Environmentally Preferable
Género
Aplicaciones de vestimenta