ESD safety shoes, lace-up, Wellmax, Milow Low
Proveedor: ELTEN
729440/40 729440/41 729440/42 729440/43 729440/44 729440/45 729440/46 729440/47 729440/48
114-6990EA 101.9 EUR
114-6990 114-6991 114-6992 114-6993 114-6994 114-6995 114-6996 114-6997 114-6998
ESD safety shoes, lace-up, Wellmax, Milow Low
Zapatos y botas
Lightweight, comfortable ESD safety shoes made from hydrophobised microfibre and hydrophobised textile upper, full padded bellows tongue and collar plus a dual-density TPU/PU sole (Wellmaxx).
  • Climate-regulating, breathable textile lining
  • Replaceable insole; orthopaedic insoles possible
  • Full length inlay sole (ESD Pro), black
  • Soft fleece ESD insole
  • Cushioning midsole made of Infinergy® by BASF rebounds back extremely well when under pressure giving the energy back to the wearer
  • Protective composite toecap (200 J)
Ideal for use in areas where exposureMore Product Information