Tychem® 6000 AL TF630 00 Ventilated Coveralls
Proveedor: DuPont
TF0630TGY00/2X TF0630TGY00/3X TF0630TGY00/L TF0630TGY00/M TF0630TGY00/S TF0630TGY00/XL
114-9631EA 440 EUR
114-9631 114-9632 114-9629 114-9628 114-9627 114-9630
Tychem® 6000 AL TF630 00 Ventilated Coveralls
Prendas desechables
Air-fed coverall with cuff elastication and integrated dissipative socks.
  • Ventilated coverall with stitched and overtaped seams
  • Fully enclosed over hood with clear front and crown see-through visor
  • Front closure with double flap hook and loop, single zipper closing to center, horizontal opening
  • Three exhaust valves; two at hips, one in hood back
  • Air distribution made of Tychem 6000 fabric
  • Emergency egress
  • Cuff elastication and elastic thumb loop on inner sleeve opening
  • Integrated dissipative socks
Tychem® 6000 AL garments are made from a lightweightMore Product Information