Toallitas para limpieza de salas, AlphaWipe®
Proveedor: Texwipe
TX1004 TX1013 TX1009
115-0496EA 130 EUR
115-0496 115-0454 115-1949
Toallitas para limpieza de salas, AlphaWipe®
Toallitas Toallitas para salas limpias Seco
These premium heavy-weight all-purpose wipes are made from 100% continuous filament polyester in a double-knit, no run, interlocked pattern. Soft and non abrasive, they are ideal for critical environments where contamination control is essential.
  • Ultra-low particulate and fibre levels
  • Excellent for sensitive surfaces
  • Superior strength guards against abrasion under rigorous use
Suitable for use in Class 10 to 100000, ISO 4 to 8 cleanrooms.
Color: blanco.
Precaución: Data accurate at time of publishing, latest test results and test methods can beMore Product Information